Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminophenol, 99%

Catalog No. AC146100010
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle

CAS: 591-27-5 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-27-5
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00007786
InChI Key CWLKGDAVCFYWJK-UHFFFAOYSA-N
Synonym m-aminophenol, m-hydroxyaniline, 3-hydroxyaniline, phenol, 3-amino, 1-amino-3-hydroxybenzene, 3-amino-1-hydroxybenzene, fouramine eg, futramine eg, fourrine eg, pelagol eg
PubChem CID 11568
ChEBI CHEBI:28924
IUPAC Name 3-aminophenol
SMILES NC1=CC=CC(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 121.0°C to 124.0°C
Color Gray or Pink to White
Boiling Point 164°C (15.0 hPa)
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4OH
Quantity 1 kg
Beilstein 13, 401
Merck Index 15, 456
Solubility Information Solubility in water: 35g/L (20°C). Other solubilities: freely soluble in hot water, freely soluble in alcohol, ether and amyl alcohol, slightly soluble in benzene, very slightly soluble in petroleum ether
Formula Weight 109.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminophenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 209-711-2
RTECSNumber SJ4900000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.