Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzaldehyde, 98%

Catalog No. AC147090025
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle

CAS: 653-37-2 | C7HF5O | 196.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-37-2
Molecular Formula C7HF5O
Molecular Weight (g/mol) 196.08
MDL Number MFCD00003303
InChI Key QJXCFMJTJYCLFG-UHFFFAOYSA-N
Synonym pentafluorobenzaldehyde, perfluorobenzaldehyde, benzaldehyde, pentafluoro, benzaldehyde, 2,3,4,5,6-pentafluoro, unii-s9yie3jz4k, pentafluor-benzaldehyd, s9yie3jz4k, pubchem4253, pentafluoro-benzaldehyde, acmc-209nqh
PubChem CID 69558
IUPAC Name 2,3,4,5,6-pentafluorobenzaldehyde
SMILES C(=O)C1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Melting Point 20°C
Color Yellow
Density 1.5880g/mL
Boiling Point 164°C to 166°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4490 to 1.451
Linear Formula C6F5CHO
Quantity 2.5 g
Specific Gravity 1.588
Formula Weight 196.08
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Pentafluorobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
Precautionary Statement GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-502-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.