Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminophenylacetylene, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
54060-30-9
C8H7N
117.15
MFCD00014779
NNKQLUVBPJEUOR-UHFFFAOYSA-N
3-aminophenylacetylene,benzenamine, 3-ethynyl,m-aminophenylacetylene,3-ethynylbenzenamine,3-ethynylphenylamine,3-ethynylaniline,m-aminophenyl acetylene,3-aminophenyl acetylene,m-ethynylaniline,3-ethynyl-aniline
104682
3-ethynylaniline
C#CC1=CC(=CC=C1)N
54060-30-9
C8H7N
117.15
MFCD00014779
NNKQLUVBPJEUOR-UHFFFAOYSA-N
3-aminophenylacetylene,benzenamine, 3-ethynyl,m-aminophenylacetylene,3-ethynylbenzenamine,3-ethynylphenylamine,3-ethynylaniline,m-aminophenyl acetylene,3-aminophenyl acetylene,m-ethynylaniline,3-ethynyl-aniline
104682
3-ethynylaniline
C#CC1=CC(=CC=C1)N
Refractive Index 1.6140 to 1.616
Boiling Point 92°C to 93°C (2.0mmHg)
Density 1.0400g/mL
Flash Point 59°C
Specific Gravity 1.04
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Yellow to Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula H2N(C6H4)CCH
Formula Weight 117.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Aminophenylacetylene, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 258-944-6