Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminophenylboronic Acid Hemisulfate, TCI America™
SDP

Catalog No. A177425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1774-25G 25 g
A1774-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1774-25G Supplier TCI America Supplier No. A177425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66472-86-4
Molecular Formula C12H18B2N2O8S
Molecular Weight (g/mol) 371.96
MDL Number MFCD00013111
InChI Key UKTAURVTSWDIQR-UHFFFAOYSA-N
PubChem CID 16211139
IUPAC Name bis((3-aminophenyl)boronic acid); sulfuric acid
SMILES OS(O)(=O)=O.NC1=CC=CC(=C1)B(O)O.NC1=CC=CC(=C1)B(O)O

Specifications

Quantity 25 g
Synonym 3-aminophenyl boronic acid sulfate 2:1, 3-aminobenzeneboronic acid hemisulfate salt, 3-aminophenylboronic acid hemisulfate, 3-aminobenzeneboronic acid hemisulfate, 3-aminophenylboronic acid hemisulphate
Formula Weight 185.98
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.