Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminophenylboronic acid monohydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
206658-89-1
C6H8BNO2·H2O
154.97
MFCD00149554
XAEOVQODHLLNKX-UHFFFAOYSA-N
3-aminophenylboronic acid monohydrate,3-aminophenyl boronic acid hydrate,3-aminophenylboronic acid hydrate,3-aminobenzeneboronic acid monohydrate,m-aminophenylboronic acid hydrate,boronic acid, 3-aminophenyl-, monohydrate,3-aminobenzeneboronic acid hydrate,pubchem4016,acmc-1cfgg,3-boronoaniline monohydrate
14389423
(3-aminophenyl)boronic acid;hydrate
B(C1=CC(=CC=C1)N)(O)O.O
206658-89-1
C6H8BNO2·H2O
154.97
MFCD00149554
XAEOVQODHLLNKX-UHFFFAOYSA-N
3-aminophenylboronic acid monohydrate,3-aminophenyl boronic acid hydrate,3-aminophenylboronic acid hydrate,3-aminobenzeneboronic acid monohydrate,m-aminophenylboronic acid hydrate,boronic acid, 3-aminophenyl-, monohydrate,3-aminobenzeneboronic acid hydrate,pubchem4016,acmc-1cfgg,3-boronoaniline monohydrate
14389423
(3-aminophenyl)boronic acid;hydrate
B(C1=CC(=CC=C1)N)(O)O.O
Melting Point 94°C to 97°C
Color Beige to Brown
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula NH2C6H4B(OH)2·H2O
Solubility Information Solubility in water: soluble
Formula Weight 154.97
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material 3-Aminophenylboronic acid monohydrate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning