Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-aminophthalic acid, 95%

Catalog No. AC375750050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 5434-20-8 | C8H7NO4 | 181.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5434-20-8
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00075053
InChI Key WGLQHUKCXBXUDV-UHFFFAOYSA-N
Synonym 3-amino-1,2-benzenedicarboxylic acid, phthalic acid, 3-amino, 3-aminophthalicacid, 1,2-benzenedicarboxylic acid, 3-amino, 3-amino-phthalic acid, unii-7xv0v19zdg, 3-aminobenzene-1,2-dicarboxylic acid, 1,2-benzenedicarboxylic acid, amino, 7xv0v19zdg, aminophthalic acid
PubChem CID 79490
IUPAC Name 3-aminophthalic acid
SMILES C1=CC(=C(C(=C1)N)C(=O)O)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 170.0°C to 180.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula NH2C6H3(CO2H)2
Quantity 5 g
Formula Weight 181.15
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminophthalic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
EINECSNumber 226-595-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.