Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopropionitrile, 98%, stabilized, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
151-18-8
C3H6N2
70.09
AGSPXMVUFBBBMO-UHFFFAOYSA-N
3-aminopropionitrile,2-cyanoethylamine,aminopropionitrile,beta-aminopropionitrile,bapn,3-aminopropiononitrile,beta-cyanoethylamine,propanenitrile, 3-amino,beta-alaninenitrile,propionitrile, 3-amino
1647
CHEBI:27413
3-aminopropanenitrile
C(CN)C#N
151-18-8
C3H6N2
70.09
AGSPXMVUFBBBMO-UHFFFAOYSA-N
3-aminopropionitrile,2-cyanoethylamine,aminopropionitrile,beta-aminopropionitrile,bapn,3-aminopropiononitrile,beta-cyanoethylamine,propanenitrile, 3-amino,beta-alaninenitrile,propionitrile, 3-amino
1647
CHEBI:27413
3-aminopropanenitrile
C(CN)C#N
Specific Gravity 0.9584
Refractive Index 1.4365 to 1.4395
Stabilizer potassium carbonate
Packaging Glass bottle
Quantity 25g
Boiling Point 185.0°C
Density 0.9584g/mL
Flash Point 85°C
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Merck Index 15,465
Solubility Information Solubility in water: miscible.
Formula Weight 70.09
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 3-Aminopropionitrile, Stabilized
Show More Show Less

GHS H Statement
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove

GHS Signal Word: Danger

EINECSNumber : 205-786-