Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Aminopropyl)Triethoxysilane, Sigma-Aldrich

Catalog No. 01258813 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-813 100 mL
01-259-009 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-813 Supplier MilliporeSigma Supelco Supplier No. A3648100ML
Add to Cart
Add to Cart
  • (3-Aminopropyl)triethoxysilane has been used for the preparation of cadherin surfaces on glass slides.
  • It has been used for the preparation of RGD (Arg-Gly-Asp)-treated silicone membranes.
  • It has been used for the preparation of the avidin-biotin coverslips.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 919-30-2

Specifications

For Use With Deactivating and Coating Chromatographic Supports
Standard Concentration 98 to 100%
Quantity 100 mL
Molecular Weight (g/mol) 221.37 g/mol, 221.37
Molecular Formula C9H23NO3Si
InChI Key WYTZZXDRDKSJID-UHFFFAOYSA-N
SMILES CCO[Si](CCCN)(OCC)OCC
IUPAC Name (3-aminopropyl)triethoxysilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.