Learn More
(3-Aminopropyl)trimethoxysilane, 97%
Description
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).
Solubility
Miscible with water.
Notes
Moisture sensitive. Incompatible withstrong acids, strong oxidizing agents, water, bases, alcohols and peroxides.

Specifications
Specifications
Density | 1.02 |
Boiling Point | 194°C |
Flash Point | 90°C (194°F) |
Odor | Amine-like |
Refractive Index | 1.424 |
Linear Formula | H2N(CH2)3Si(OCH3)3 |
Quantity | 500 g |
UN Number | UN2735 |
Beilstein | 2038118 |
Sensitivity | Air and moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H227-H315-H318 |
Hazard Statement | GHS H Statement H314-H318-H227 Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
Precautionary Statement | P210-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 237-511-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.