Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopyridazine, 97%

Catalog No. AAH6485906
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 5469-70-5 | C4H5N3 | 95.105 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5469-70-5
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.105
MDL Number MFCD01529869
InChI Key LETVJWLLIMJADE-UHFFFAOYSA-N
Synonym 3-aminopyridazine, 3-pyridazinamine, aminopyridazine, pyridazin-3-ylamine, 3-amino-1,2-diazine, pyridazine-3-ylamine, pyridazineamine, pyridazyl amine, imino-pyridazine, 3-pyridazinamin
PubChem CID 230373
IUPAC Name pyridazin-3-amine
SMILES C1=CC(=NN=C1)N

Specifications

Melting Point 168°C to 172°C
Color White to Yellow
Quantity 5 g
Beilstein 107024
Formula Weight 95.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Aminopyridazine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.