Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Aminopyridazine, 97%

Click to view available options
Quantity:
1g
5g
25g
5469-70-5
C4H5N3
95.105
MFCD01529869
LETVJWLLIMJADE-UHFFFAOYSA-N
3-aminopyridazine,3-pyridazinamine,aminopyridazine,pyridazin-3-ylamine,3-amino-1,2-diazine,pyridazine-3-ylamine,pyridazineamine,pyridazyl amine,imino-pyridazine,3-pyridazinamin
230373
pyridazin-3-amine
C1=CC(=NN=C1)N
5469-70-5
C4H5N3
95.105
MFCD01529869
LETVJWLLIMJADE-UHFFFAOYSA-N
3-aminopyridazine,3-pyridazinamine,aminopyridazine,pyridazin-3-ylamine,3-amino-1,2-diazine,pyridazine-3-ylamine,pyridazineamine,pyridazyl amine,imino-pyridazine,3-pyridazinamin
230373
pyridazin-3-amine
C1=CC(=NN=C1)N
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Melting Point 168°C to 172°C
Quantity 1g
Color White to Yellow
Beilstein 107024
Formula Weight 95.1
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Aminopyridazine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning