Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopyridine-2-carboxylic acid, 97%

Catalog No. AAH6486106
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAH6486106

Supplier: Thermo Scientific Chemicals H6486106

Only null left
Add to Cart

CAS: 1462-86-8 | C6H6N2O2 | 138.126 g/mol

Employed as an important intermediate for syntheses material and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for syntheses material and pharmaceutical.

Solubility
Soluble in acetone, ethanol and methanol.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1462-86-8
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00090153
InChI Key BOOMHTFCWOJWFO-UHFFFAOYSA-N
Synonym 3-aminopicolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3-amino, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-2-carboxypyridine, 2-pyridinecarboxylicacid, 3-amino, pubchem2015, 3-aminopyridine-2-carboxylic acid, tech.
PubChem CID 73836
IUPAC Name 3-aminopyridine-2-carboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)N

Specifications

Melting Point 218°C to 222°C
Color Brown
Quantity 5 g
Beilstein 3600
Solubility Information Soluble in acetone,ethanol and methanol.
Formula Weight 138.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Aminopyridine-2-carboxylic acid
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 215-971-8
RTECSNumber US5175000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.