Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopyridine, 99%

Catalog No. AAA132160B
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA132160B 1000 g
AAA1321618 50 g
AAA1321630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA132160B Supplier Thermo Scientific Chemicals Supplier No. A132160B
Add to Cart

CAS: 462-08-8 | C5H6N2 | 94.12 g/mol

3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.

Solubility
It is soluble in water as well as soluble in alcohol, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under ordinary conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-08-8
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.12
MDL Number MFCD00006400
InChI Key CUYKNJBYIJFRCU-UHFFFAOYSA-N
Synonym 3-aminopyridine, 3-pyridinamine, 3-pyridylamine, pyridin-3-ylamine, m-aminopyridine, amino-3 pyridine, pyridine, 3-amino, 3-amino pyridine, beta-aminopyridine, amino-3-pyridine
PubChem CID 10009
IUPAC Name pyridin-3-amine
SMILES NC1=CC=CN=C1

Specifications

Melting Point 58°C to 63°C
pH 10
Boiling Point 250°C to 252°C
Flash Point 88°C (190°F)
Odor Unpleasant
Quantity 1000 g
UN Number UN2671
Beilstein 105692
Sensitivity Hygroscopic
Merck Index 14,473
Solubility Information It is soluble in water as well as soluble in alcohol,benzene.
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material 3-Aminopyridine
Show More Show Less
Hazard Category H300-H311-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES
EINECSNumber 207-322-2
RTECSNumber US1650000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.