Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Azabicyclo[3.3.0]octane hydrochloride, 99%, Thermo Scientific™

Catalog No. AC457700050
Click to view available options
Quantity:
5 g
25 g
100 g

Chemical Identifiers

CAS 112626-50-3
Molecular Formula C7H13N·ClH
Molecular Weight (g/mol) 147.65
InChI Key HVZRRRPCVOJOLJ-UHFFFAOYSA-N
Synonym 3-azabicyclo 3.3.0 octane hydrochloride, octahydrocyclopenta c pyrrole hydrochloride, 3-azabicyclo 3.3.0 octane hcl, 3-azabicyclo 3,3,0 octane hcl, 3-azabicyclo 3.3.0 octanehydrochloride, 1,2,3,3a,4,5,6,6a-octahydrocyclopenta c pyrrole hydrochloride, pubchem9779, cis-octahydro-cyclopenta c pyrrole hydrochloride, c7h14ncl, 3-azabicyclo 3.3.0 octane hydrochloride;
PubChem CID 21572123
IUPAC Name 1,2,3,3a,4,5,6,6a-octahydrocyclopenta[c]pyrrole;hydrochloride
SMILES C1CC2CNCC2C1.Cl

Specifications

Melting Point 116.0°C to 118.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5 to max. 101.5% min. (Argentometry)
Quantity 5 g
Formula Weight 147.65
Percent Purity 99%
Chemical Name or Material 3-Azabicyclo[3.3.0]octane hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.