Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Azido-1-Propylamine, PROMO, Thermo Scientific Chemicals

Catalog No. J6566803PM
Click to view available options
Quantity:
1 g

Azido-containing reagent with carbonyl and carboxyl reactivity

Applications

Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.

Solubility

Soluble in DMSO, DMF, DCM, chloroform, THF, and water.Miscible with dimethylsulfoxide, dimethyl formamide, dichloromethane, tetrahydrofuran, chloroform and water.

Notes

Incompatible with strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88192-19-2
Molecular Formula C3H8N4
Molecular Weight (g/mol) 100.13 g/mol
MDL Number MFCD11046568
InChI Key OYBOVXXFJYJYPC-UHFFFAOYSA-N
Synonym 3-azidopropylamine, 3-azido-1-propanamine, 1-propanamine, 3-azido, 3-azido-propylamine, 1-azido-3-aminopropane, 1-amino-3-azidopropane, 3-aminopropyl azide, 3-azido-1-propylamine, 3-azidopropyl-1-amine
PubChem CID 150110
IUPAC Name 3-azidopropan-1-amine
SMILES C(CN)CN=[N+]=[N-]

Specifications

Color Yellow
Flash Point 60°C (140°F)
Quantity 1 g
UN Number UN1993
Solubility Information Soluble in DMSO, DMF, DCM, chloroform, THF, and water
Physical Form Liquid
Chemical Name or Material 3-Azido-1-propylamine
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.