Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trifluoroacetic acid-d, for NMR, packaged in 0.75 ml ampoules, 99.5 atom % D
Click to view available options
Quantity:
7.5 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Specific Gravity | 1.5 |
| Quantity | 7.5 mL |
| Density | 1.5000 g/mL |
| Melting Point | -15.0°C |
| Boiling Point | 75.0°C |
| Linear Formula | CF3CO2D |
| Solubility Information | Solubility aq. soln.: soluble. Other solubilities: miscible with ether, acetone, ethanol, benzene,, ccl4, hexane, methanol, xylene and petroleumether |
| Formula Weight | 115.03 |
| Grade | NMR |
| Chemical Name or Material | Trifluoroacetic acid-d |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful to aquatic life with long lasting effects. Harmful if inhaled. Causes severe skin burns and eye damage. May be corrosive to metals. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. |
| EINECSNumber | 209-961-2 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction