Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzyloxazolidine 98.0+%, TCI America™
SDP

Catalog No. B13391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13391G 1 g
B13395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13391G Supplier TCI America Supplier No. B13391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13657-16-4
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00059028
InChI Key FQNODHUYZYLTPN-UHFFFAOYSA-N
PubChem CID 319258
IUPAC Name 3-benzyl-1,3-oxazolidine
SMILES C1COCN1CC2=CC=CC=C2

Specifications

Color Yellow
Boiling Point 127°C
Quantity 1 g
Formula Weight 163.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Benzyloxazolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.