Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzyloxy-5-bromopyridine 98.0+%, TCI America™
SDP

Catalog No. B47751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47751G 1 g
B47755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47751G Supplier TCI America Supplier No. B47751G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 130722-95-1
Molecular Formula C12H10BrNO
Molecular Weight (g/mol) 264.122
MDL Number MFCD07375004
InChI Key YSHKYZAWTWKQKK-UHFFFAOYSA-N
PubChem CID 15389198
IUPAC Name 3-bromo-5-phenylmethoxypyridine
SMILES C1=CC=C(C=C1)COC2=CC(=CN=C2)Br

Specifications

Melting Point 71°C
Color Yellow
Quantity 1 g
Formula Weight 264.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzyloxy-5-bromopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.