Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzyloxyacetophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
34068-01-4
C15H14O2
226.27
MFCD00026221
FGQMEAWGAUALJQ-UHFFFAOYSA-N
1-3-benzyloxy phenyl ethanone,3-benzyloxyacetophenone,3'-benzyloxyacetophenone,1-3-benzyloxy phenyl ethan-1-one,1-3-phenylmethoxy phenyl ethan-1-one,3'-benzyloxy acetophenone,ethanone, 1-3-phenylmethoxy phenyl,1-acetyl-3-phenylmethoxy benzene
98689
1-(3-phenylmethoxyphenyl)ethanone
CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2
34068-01-4
C15H14O2
226.27
MFCD00026221
FGQMEAWGAUALJQ-UHFFFAOYSA-N
1-3-benzyloxy phenyl ethanone,3-benzyloxyacetophenone,3'-benzyloxyacetophenone,1-3-benzyloxy phenyl ethan-1-one,1-3-phenylmethoxy phenyl ethan-1-one,3'-benzyloxy acetophenone,ethanone, 1-3-phenylmethoxy phenyl,1-acetyl-3-phenylmethoxy benzene
98689
1-(3-phenylmethoxyphenyl)ethanone
CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2
Color Yellow
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Beilstein 08, IV, 335
Formula Weight 226.27
Physical Form Low Melting Solid
Percent Purity 97%
Chemical Name or Material 3-Benzyloxyacetophenone, 97%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 251-816-0

TSCA : TSCA