Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzyloxyphenyl isothiocyanate, 97%

Catalog No. AAL1924203
Click to view available options
Quantity:
250 mg
1 g

CAS: 206559-36-6 | C14H11NOS | 241.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206559-36-6
Molecular Formula C14H11NOS
Molecular Weight (g/mol) 241.31
MDL Number MFCD00060374
InChI Key QJLFNDXMZQJMDL-UHFFFAOYSA-N
Synonym 3-benzyloxyphenyl isothiocyanate, 3-benzyloxyphenylisothiocyanate, 1-benzyloxy-3-isothiocyanatobenzene, acmc-20an9f, 3-phenylmethoxy benzenisothiocyanate, 1-benzyloxy-3-isothiocyanatobenzene #, 1-isothiocyanato-3-phenylmethoxy-benzene, benzene, 1-isothiocyanato-3-phenylmethoxy, benzene, 1-isothiocyanato-3-phenylmethoxy-9ci
PubChem CID 561446
IUPAC Name 1-isothiocyanato-3-phenylmethoxybenzene
SMILES S=C=NC1=CC(OCC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 49°C to 51°C
Quantity 1 g
UN Number UN2923
Sensitivity Moisture sensitive
Formula Weight 241.31
Percent Purity 97%
Chemical Name or Material 3-Benzyloxyphenyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.