Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzyloxypyrrolidine, 96%

Catalog No. AAH5076903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5076903 1 g
AAH50769MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5076903 Supplier Thermo Scientific Chemicals Supplier No. H5076903
Add to Cart

CAS: 420137-14-0 | C11H15NO | 177.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 420137-14-0
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD08060975
InChI Key CWBMYKUPMLRKQK-UHFFFAOYSA-N
Synonym 3-benzyloxy pyrrolidine, 3-benzyloxypyrrolidine, 3-benzyloxy-pyrrolidine, 3-phenylmethoxy pyrrolidine, benzyl pyrrolidin-3-yl ether, pyrrolidin-3-yl oxy methyl benzene
PubChem CID 14497353
IUPAC Name 3-phenylmethoxypyrrolidine
SMILES C1CNCC1OCC2=CC=CC=C2

Specifications

Quantity 1 g
Formula Weight 177.25
Percent Purity 96%
Chemical Name or Material 3-Benzyloxypyrrolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.