Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-1-(phenylsulfonyl)indole, 97%

Click to view available options
Quantity:
1g
5g
99655-68-2
C14H10BrNO2S
336.203
MFCD02681986
MXPFKHHDXIJNDX-UHFFFAOYSA-N
3-bromo-1-phenylsulfonyl-1h-indole,1-benzenesulfonyl-3-bromoindole,3-bromo-1-phenylsulfonyl indole,1-benzenesulfonyl-3-bromo-1h-indole,1-phenylsulfonyl-3-bromoindole,1-benzenesulphonyl-3-bromo-1h-indole,3-bromo-1-phenylsulphonyl-1h-indole,1-benzenesulfonyl-3-bromo-1h-indole,3-bromo-1-benzenesulphonylindole,1h-indole, 3-bromo-1-phenylsulfonyl
2776215
1-(benzenesulfonyl)-3-bromoindole
C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)Br
99655-68-2
C14H10BrNO2S
336.203
MFCD02681986
MXPFKHHDXIJNDX-UHFFFAOYSA-N
3-bromo-1-phenylsulfonyl-1h-indole,1-benzenesulfonyl-3-bromoindole,3-bromo-1-phenylsulfonyl indole,1-benzenesulfonyl-3-bromo-1h-indole,1-phenylsulfonyl-3-bromoindole,1-benzenesulphonyl-3-bromo-1h-indole,3-bromo-1-phenylsulphonyl-1h-indole,1-benzenesulfonyl-3-bromo-1h-indole,3-bromo-1-benzenesulphonylindole,1h-indole, 3-bromo-1-phenylsulfonyl
2776215
1-(benzenesulfonyl)-3-bromoindole
C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)Br
Quantity 5g
Melting Point 122°C to 126°C
Beilstein 3553533
Formula Weight 336.2
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Bromo-1-(phenylsulfonyl)indole

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning