Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-1-propanol, 97%

Click to view available options
Quantity:
5g
25g
100g
627-18-9
C3H7BrO
138.992
MFCD00002942
RQFUZUMFPRMVDX-UHFFFAOYSA-N
3-bromo-1-propanol,trimethylene bromohydrin,1-propanol, 3-bromo,3-bromopropanol,3-hydroxypropyl bromide,1-bromo-3-propanol,3-bromo-propan-1-ol,ccris 5979,1-bromo-3-hydroxypropane,unii-3t4axh68fh
12308
3-bromopropan-1-ol
C(CO)CBr
627-18-9
C3H7BrO
138.992
MFCD00002942
RQFUZUMFPRMVDX-UHFFFAOYSA-N
3-bromo-1-propanol,trimethylene bromohydrin,1-propanol, 3-bromo,3-bromopropanol,3-hydroxypropyl bromide,1-bromo-3-propanol,3-bromo-propan-1-ol,ccris 5979,1-bromo-3-hydroxypropane,unii-3t4axh68fh
12308
3-bromopropan-1-ol
C(CO)CBr
Applications
3-Bromo-1-propanol is used in the synthesis of fluorescent halide-sensitive quinolinium dyes, chiral, quaternary prolines through cyclization of quaternary amino acids and molten salt-polymers. It is utilized for the study of micellar media and in microemulsions based on cationic or a nonionic surfactant by reacting with phenols. It acts as a grafting agent in the synthesis of recyclable reagents and an intermediate in the synthesis of organic molecules.

Solubility
Miscible with water. Slightly miscible with ethanol and ether.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Flash Point 93°C (199°F)
Boiling Point 71°C to 73°C (11mmHg)
Quantity 100g
Linear Formula BrCH2CH2CH2OH
Beilstein 969160
Formula Weight 138.99
Percent Purity ≥97%
Chemical Name or Material 3-Bromo-1-propanol

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P261-P280g-P305+P351+P338
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : UA7385000

TSCA : TSCA

Recommended Storage : 2° to 8°C