Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-1-propanol, 95%

Catalog No. p-7026699
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 627-18-9 | C3H7BrO | 138.99 g/mol

3-Bromo-1-propanol is used in the synthesis of fluorescent halide-sensitive quinolinium dyes, chiral, quaternary prolines through cyclization of quaternary amino acids and molten salt-polymers. It is utilized for the study of micellar media and in microemulsions based on cationic or a nonionic surfactant by reacting with phenols. It acts as a grafting agent in the synthesis of recyclable reagents and an intermediate in the synthesis of organic molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-1-propanol is used in the synthesis of fluorescent halide-sensitive quinolinium dyes, chiral, quaternary prolines through cyclization of quaternary amino acids and molten salt-polymers. It is utilized for the study of micellar media and in microemulsions based on cationic or a nonionic surfactant by reacting with phenols. It acts as a grafting agent in the synthesis of recyclable reagents and an intermediate in the synthesis of organic molecules.

Solubility
Miscible with water. Slightly miscible with ethanol and ether.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-18-9
Molecular Formula C3H7BrO
Molecular Weight (g/mol) 138.99
MDL Number MFCD00002942
InChI Key RQFUZUMFPRMVDX-UHFFFAOYSA-N
Synonym 3-bromo-1-propanol, trimethylene bromohydrin, 1-propanol, 3-bromo, 3-bromopropanol, 3-hydroxypropyl bromide, 1-bromo-3-propanol, 3-bromo-propan-1-ol, ccris 5979, 1-bromo-3-hydroxypropane, unii-3t4axh68fh
PubChem CID 12308
IUPAC Name 3-bromopropan-1-ol
SMILES OCCCBr

Specifications

Density 1.59
Boiling Point 71°C to 73°C (11 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.4885
Linear Formula BrCH2CH2CH2OH
Quantity 5 g
Beilstein 969160
Solubility Information Miscible with water. Slightly miscible with ethanol and ether.
Formula Weight 138.99
Percent Purity ≥97%
Chemical Name or Material 3-Bromo-1-propanol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-986-6
RTECSNumber UA7385000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.