Alfa Aesar™ 3-Bromo-1-propanol, 97%

Manufacturer: Alfa Aesar™ B2285706

Quantity 5g

 View more versions of this product

Catalog No. AAB2285706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 627-18-9
    Molecular Formula C3H7BrO
    Linear Formula BrCH2CH2CH2OH
    Formula Weight 138.99
    MDL Number MFCD00002942
    Beilstein 969160
    Synonym Trimethylene bromohydrin; 1-Bromo-3-hydroxypropane
    Chemical Name or Material 3-Bromo-1-propanol

    Specifications

    Formula Weight 138.99
    TSCA TSCA
    Boiling Point 71° to 73°C (11mmHg)
    Quantity 5g
    Flash Point 93°C (199°F)
    Molecular Formula C3H7BrO
    Linear Formula BrCH2CH2CH2OH
    MDL Number MFCD00002942
    Beilstein 969160
    Synonym Trimethylene bromohydrin; 1-Bromo-3-hydroxypropane
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Recommended Storage 2° to 8°C
    RTECS Number UA7385000
    CAS 627-18-9
    Health Hazard 3 GHS P Statement
    P261-P280g-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 3-Bromo-1-propanol
    Assay ≥97%

    Description

    Applications
    3-Bromo-1-propanol is used in the synthesis of fluorescent halide-sensitive quinolinium dyes, chiral, quaternary prolines through cyclization of quaternary amino acids and molten salt-polymers. It is utilized for the study of micellar media and in microemulsions based on cationic or a nonionic surfactant by reacting with phenols. It acts as a grafting agent in the synthesis of recyclable reagents and an intermediate in the synthesis of organic molecules.

    Solubility
    Miscible with water. Slightly miscible with ethanol and ether.

    Notes
    Store in cool place. Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P261-P280g-P305+P351+P338
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit