Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2,5-diaminopyridine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
896160-69-3
C5H6BrN3
188
MFCD03427651
YJENCMIONVUOCW-UHFFFAOYSA-N
2,5-diamino-3-bromopyridine,3-bromo-2,5-diaminopyridine,2,5-diamino-3-bormopyridine,2,5-pyridinediamine, 3-bromo,pubchem5490,acmc-1c3l9,3-bromanylpyridine-2,5-diamine,abbypharma ap-31-4166
2734413
3-bromopyridine-2,5-diamine
C1=C(C=NC(=C1Br)N)N
896160-69-3
C5H6BrN3
188
MFCD03427651
YJENCMIONVUOCW-UHFFFAOYSA-N
2,5-diamino-3-bromopyridine,3-bromo-2,5-diaminopyridine,2,5-diamino-3-bormopyridine,2,5-pyridinediamine, 3-bromo,pubchem5490,acmc-1c3l9,3-bromanylpyridine-2,5-diamine,abbypharma ap-31-4166
2734413
3-bromopyridine-2,5-diamine
C1=C(C=NC(=C1Br)N)N
Melting Point 117°C to 122°C
Packaging Glass bottle
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 188
Percent Purity 99%
Chemical Name or Material 3-Bromo-2, 5-diaminopyridine

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning