Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-chloropyridine, 97%, Thermo Scientific Chemicals

Catalog Number AAB2520614
Click to view available options
Quantity:
5 g
25 g
52200-48-3
C5H3BrClN
192.44
MFCD00234007
HDYNIWBNWMFBDO-UHFFFAOYSA-N
2-chloro-3-bromopyridine, pyridine, 3-bromo-2-chloro, 3-bromo-2-chloro-pyridine, pubchem1098, 2-chloro-bromopyridine, acmc-209kyb, 3-bromo-2-chlorolpyridine, 3-bromo-2-chloropyridine, 3-bromo-2-chloro pyridine, 2-chloro-3-bromo pyridine
693324
3-bromo-2-chloropyridine
C1=CC(=C(N=C1)Cl)Br
52200-48-3
C5H3BrClN
192.44
MFCD00234007
HDYNIWBNWMFBDO-UHFFFAOYSA-N
2-chloro-3-bromopyridine, pyridine, 3-bromo-2-chloro, 3-bromo-2-chloro-pyridine, pubchem1098, 2-chloro-bromopyridine, acmc-209kyb, 3-bromo-2-chlorolpyridine, 3-bromo-2-chloropyridine, 3-bromo-2-chloro pyridine, 2-chloro-3-bromo pyridine
693324
3-bromo-2-chloropyridine
C1=CC(=C(N=C1)Cl)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 52°C to 57°C
Boiling Point 97°C (10 mmHg)
Quantity 25 g
Beilstein 109812
Formula Weight 192.44
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-chloropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit