Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-chloropyridine, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
52200-48-3
C5H3BrClN
192.44
HDYNIWBNWMFBDO-UHFFFAOYSA-N
2-chloro-3-bromopyridine,pyridine, 3-bromo-2-chloro,3-bromo-2-chloro-pyridine,pubchem1098,2-chloro-bromopyridine,acmc-209kyb,3-bromo-2-chlorolpyridine,3-bromo-2-chloropyridine,3-bromo-2-chloro pyridine,2-chloro-3-bromo pyridine
693324
3-bromo-2-chloropyridine
C1=CC(=C(N=C1)Cl)Br
52200-48-3
C5H3BrClN
192.44
HDYNIWBNWMFBDO-UHFFFAOYSA-N
2-chloro-3-bromopyridine,pyridine, 3-bromo-2-chloro,3-bromo-2-chloro-pyridine,pubchem1098,2-chloro-bromopyridine,acmc-209kyb,3-bromo-2-chlorolpyridine,3-bromo-2-chloropyridine,3-bromo-2-chloro pyridine,2-chloro-3-bromo pyridine
693324
3-bromo-2-chloropyridine
C1=CC(=C(N=C1)Cl)Br
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 97°C (1.3 kPa)
Color White to Yellow
Melting Point 57.9°C to 58.3°C
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 192.44
Physical Form Crystalline Powder, Crystals or Flakes or Solid
Percent Purity 98%
Chemical Name or Material 3-Bromo-2-chloropyridine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Warning