Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-chlorobenzotrifluoride 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
454-78-4
C7H3BrClF3
259.45
MFCD00018093
APSISOSWYXCEQX-UHFFFAOYSA-N
2-bromo-1-chloro-4-trifluoromethyl benzene,3-bromo-4-chlorobenzotrifluoride,benzene, 2-bromo-1-chloro-4-trifluoromethyl,3-bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene,4-chloro-3-bromobenzotrifluoride,3-bromo-4-chloro-a,a,a-trifluorotoluene,pubchem4837,ksc915a8j,apsisoswyxceqx-uhfffaoysa,3-bromo-4-chloro-benzotrifluoride
67988
2-bromo-1-chloro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Br)Cl
454-78-4
C7H3BrClF3
259.45
MFCD00018093
APSISOSWYXCEQX-UHFFFAOYSA-N
2-bromo-1-chloro-4-trifluoromethyl benzene,3-bromo-4-chlorobenzotrifluoride,benzene, 2-bromo-1-chloro-4-trifluoromethyl,3-bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene,4-chloro-3-bromobenzotrifluoride,3-bromo-4-chloro-a,a,a-trifluorotoluene,pubchem4837,ksc915a8j,apsisoswyxceqx-uhfffaoysa,3-bromo-4-chloro-benzotrifluoride
67988
2-bromo-1-chloro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Br)Cl
Refractive Index 1.4980 to 1.5
Melting Point -23°C to 22°C
Boiling Point 188°C to 190°C
Density 1.7400g/mL
Specific Gravity 1.74
Flash Point 94°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Quantity 25g
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula BrC6H3(Cl)CF3
Formula Weight 259.45
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material 3-Bromo-4-chlorobenzotrifluoride, 98+%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-226-0

TSCA : TSCA