Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-hydroxybenzyl alcohol, 98%, Thermo Scientific™

Catalog No. p-7031396
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29922-56-3
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.035
MDL Number MFCD00016869
InChI Key HAFQFRJTRZBJJF-UHFFFAOYSA-N
Synonym 2-bromo-4-hydroxymethyl phenol, 3-bromo-4-hydroxybenzyl alcohol, 2-bromo-4-hydroxymethyl-phenol, benzenemethanol,3-bromo-4-hydroxy, 2-brom-4-hydroxymethyl-phenol, 2-bromo-4-hydroxymethylphenol;, 3-bromo-4-hydroxybenzenemethanol, 3-brom-4-hydroxy-benzylalkohol;, 2-bromo-4-hydroxymethyl phenol;
PubChem CID 121594
IUPAC Name 2-bromo-4-(hydroxymethyl)phenol
SMILES C1=CC(=C(C=C1CO)Br)O

Specifications

Melting Point 128°C to 132°C
Quantity 250 mg
Formula Weight 203.04
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-hydroxybenzyl alcohol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.