Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methoxybenzonitrile, 99%, Thermo Scientific™

Catalog No. AC429880010
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 117572-79-9
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.05
MDL Number MFCD00143097
InChI Key QHWZMDRKTYTPEE-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanoanisole, 3-bromo-4-methoxylbenzonitrile, 3-bromo-4-methoxybenzenecarbonitrile, 3-bromo-4-methoxy-benzonitrile, benzonitrile, 3-bromo-4-methoxy, pubchem3795, acmc-1cexj, maybridge4_001971, 3-bromo4-methoxybenzonitrile, ksc494m8l
PubChem CID 736049
IUPAC Name 3-bromo-4-methoxybenzonitrile
SMILES COC1=CC=C(C=C1Br)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 120.0°C to 121.0°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 212.05
Percent Purity 99%
Chemical Name or Material 3-Bromo-4-methoxybenzonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.