Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methylbenzoic acid, 98+%

Catalog No. AAA1149614
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 7697-26-9 | C8H7BrO2 | 215.05 g/mol

3-Bromo-4-methylbenzoic acid has been used in the synthesis of biphenyl amides, 2-benzazepine-4-acetic acid derivative, as an analog of the potent, nonpeptide GPIIb/IIIa antagonist, O-spiro C-aryl glucosides. It is also used as catalytic agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methylbenzoic acid has been used in the synthesis of biphenyl amides, 2-benzazepine-4-acetic acid derivative, as an analog of the potent, nonpeptide GPIIb/IIIa antagonist, O-spiro C-aryl glucosides. It is also used as catalytic agent and petrochemical additive.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-26-9
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00002488
InChI Key ZFJOMUKPDWNRFI-UHFFFAOYSA-N
Synonym 3-bromo-4-methylbenzoicacid, 3-bromo-p-toluic acid, benzoic acid, 3-bromo-4-methyl, 3-bromo-4-methyl-benzoic acid, bromo-p-toluic acid, pubchem3806, bromo-para-toluic acid, 3-bromo4-methylbenzoic acid, acmc-1bg95, akos bbb/219
PubChem CID 82130
IUPAC Name 3-bromo-4-methylbenzoic acid
SMILES CC1=CC=C(C=C1Br)C(O)=O

Specifications

Melting Point 206°C to 211°C
Odor Odorless
Quantity 25 g
Beilstein 1936510
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-4-methylbenzoic acid
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 231-712-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.