Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methylbenzoic acid, 98%, Thermo Scientific™

Catalog No. AC458140050
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 7697-26-9
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00002488
InChI Key ZFJOMUKPDWNRFI-UHFFFAOYSA-N
Synonym 3-bromo-4-methylbenzoicacid, 3-bromo-p-toluic acid, benzoic acid, 3-bromo-4-methyl, 3-bromo-4-methyl-benzoic acid, bromo-p-toluic acid, pubchem3806, bromo-para-toluic acid, 3-bromo4-methylbenzoic acid, acmc-1bg95, akos bbb/219
PubChem CID 82130
IUPAC Name 3-bromo-4-methylbenzoic acid
SMILES CC1=CC=C(C=C1Br)C(O)=O

Specifications

Melting Point 209.0°C to 210.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 215.05
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-methylbenzoic acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 231-712-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.