Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methylpyridine, 97%

Catalog No. p-3593280
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC392320010 1 g Glass bottle
AC392320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC392320010 Supplier Thermo Scientific Chemicals Supplier No. 392320010
Only null left
Add to Cart

CAS: 3430-22-6 | C6H6BrN | 172.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3430-22-6
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.02
MDL Number MFCD00082592
InChI Key GSQZOLXWFQQJHJ-UHFFFAOYSA-N
Synonym 3-bromo-4-picoline, 4-methyl-3-bromopyridine, pyridine, 3-bromo-4-methyl, 3-bromo-4-methyl-pyridine, 3-brom-4-methylpyridin, zlchem 494, pubchem3994, 3-bromo4-methylpyridine, 3-bromo-4-methylpyridin
PubChem CID 817630
IUPAC Name 3-bromo-4-methylpyridine
SMILES CC1=C(C=NC=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5490g/mL
Boiling Point 76.0°C (10.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5600 to 1.5620
Quantity 1 g
Specific Gravity 1.549
Formula Weight 172.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-4-methylpyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.