Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-fluorobenzylamine hydrochloride, 96%, Thermo Scientific™

Catalog No. AAH6224003
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6224003 1 g
AAH62240MD 250 mg
AAH6224006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6224003 Supplier Thermo Scientific Chemicals Supplier No. H6224003

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189924-80-8
Molecular Formula C7H8BrClFN
Molecular Weight (g/mol) 240.5
MDL Number MFCD04113608
InChI Key OFYIDRUSNSFKEG-UHFFFAOYSA-N
Synonym 3-bromo-5-fluorophenyl methanamine hydrochloride, 3-bromo-5-fluorobenzylamine hydrochloride, 3-bromo-5-fluorophenyl methylamine, chloride, 3-bromanyl-5-fluoranyl-phenyl methanamine hydrochloride, 1-3-bromo-5-fluorophenyl methanamine hydrochloride
PubChem CID 44717343
IUPAC Name (3-bromo-5-fluorophenyl)methanamine;hydrochloride
SMILES C1=C(C=C(C=C1F)Br)CN.Cl

Specifications

Quantity 1 g
Formula Weight 240.5
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Bromo-5-fluorobenzylamine hydrochloride
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.