Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-hydroxypyridine, 95%, Thermo Scientific™

Catalog No. aah27256md
Click to view available options
Quantity:
250 mg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the total synthesis of (+-)-tabersonine.

Solubility
Slightly soluble in water. Soluble in chloroform and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74115-13-2
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 174.00
MDL Number MFCD00661305
InChI Key VNYBIBSZZDAEOK-UHFFFAOYSA-N
Synonym 3-bromo-5-hydroxypyridine, 5-bromo-3-pyridinol, 5-bromo-3-hydroxypyridine, 5-bromo-pyridin-3-ol, 3-pyridinol, 5-bromo, 3-bromo-5-pyridinol, 3-hydroxy-5-bromopyridine, 5-bromo-3-hydroxy-pyridine, pyridine, 3-ol-5-bromo, 5-bromopyrid-3-ol
PubChem CID 599529
IUPAC Name 5-bromopyridin-3-ol
SMILES OC1=CN=CC(Br)=C1

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water. Soluble in chloroform and ethyl acetate.
Formula Weight 174
Percent Purity 95%
Chemical Name or Material 3-Bromo-5-hydroxypyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.