Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-5-methylbenzotrifluoride, 99%, Thermo Scientific™

Catalog No. p-7031092
Click to view available options
Quantity:
5 g

Catalog No. AAH3204006

Supplier: Thermo Scientific Chemicals H3204006

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86845-28-5
Molecular Formula C8H6BrF3
Molecular Weight (g/mol) 239.04
MDL Number MFCD01631585
InChI Key ZDAIYLXHTGIUGY-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl toluene, 1-bromo-3-methyl-5-trifluoromethyl benzene, 3-bromo-5-methylbenzotrifluoride, 1-bromo-3-methyl-5-trifluoromethyl-benzene, 3-methyl-5-trifluoromethyl bromobenzene, 3-bromo-5-methyl-1-trifluoromethyl benzene, benzene,1-bromo-3-methyl-5-trifluoromethyl, 5-bromo-alpha,alpha,alpha-trifluoro-m-xylene, 3-bromo-5-trifluoromethyl toluene 3-bromo-5-methylbenzotrifluoride
PubChem CID 20467615
IUPAC Name 1-bromo-3-methyl-5-(trifluoromethyl)benzene
SMILES CC1=CC(=CC(Br)=C1)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 239.04
Percent Purity 99%
Chemical Name or Material 3-Bromo-5-methylbenzotrifluoride
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.