Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-9-(1-naphthyl)-9H-carbazole 97.0+%, TCI America™
SDP

Catalog No. B50501G
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 934545-83-2
Molecular Formula C22H14BrN
Molecular Weight (g/mol) 372.265
MDL Number MFCD26127423
InChI Key QTWVKYHIDBPQIL-UHFFFAOYSA-N
PubChem CID 59561149
IUPAC Name 3-bromo-9-naphthalen-1-ylcarbazole
SMILES C1=CC=C2C(=C1)C=CC=C2N3C4=C(C=C(C=C4)Br)C5=CC=CC=C53

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 372.27
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9-(1-naphthyl)-9H-carbazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.