Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-9-ethylcarbazole 98.0+%, TCI America™
SDP

Catalog No. B34591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34591G 1 g
B34595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34591G Supplier TCI America Supplier No. B34591G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 57102-97-3
Molecular Formula C14H12BrN
Molecular Weight (g/mol) 274.161
MDL Number MFCD00092392
InChI Key PPHYYUDMDADQPF-UHFFFAOYSA-N
PubChem CID 621080
IUPAC Name 3-bromo-9-ethylcarbazole
SMILES CCN1C2=C(C=C(C=C2)Br)C3=CC=CC=C31

Specifications

Melting Point 84°C
Color White
Quantity 1 g
Formula Weight 274.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-9-ethylcarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.