Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-o-xylene, 99%

Catalog No. AAA1588914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1588914 25 g
AAA1588906 5 g
AAA1588922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1588914 Supplier Thermo Scientific Chemicals Supplier No. A1588914
Add to Cart

CAS: 576-23-8 | C8H9Br | 185.06 g/mol

3-Bromo-o-xylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-o-xylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-23-8
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000069
InChI Key WLPXNBYWDDYJTN-UHFFFAOYSA-N
Synonym 3-bromo-o-xylene, 3-bromo-1,2-dimethylbenzene, 2,3-dimethylbromobenzene, 2,3-dimethyl bromobenzene, benzene, bromodimethyl, 1-bromo-2,3-dimethyl-benzene, benzene, 1-bromo-2,3-dimethyl, bromoxilene, xylylbromid, bromo-o-xylene
PubChem CID 68472
IUPAC Name 1-bromo-2,3-dimethylbenzene
SMILES CC1=CC=CC(Br)=C1C

Specifications

Density 1.365
Quantity 25 g
Refractive Index 1.56
Beilstein 2077312
Boiling Point 212°C to 214°C
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 185.07
Percent Purity 99%
Chemical Name or Material 3-Bromo-o-xylene
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 209-398-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.