Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobenzoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass Bottle
Plastic bottle
585-76-2
C7H5BrO2
201.02
MFCD00002487
VOIZNVUXCQLQHS-UHFFFAOYSA-N
m-bromobenzoic acid,3-brombenzoic acid,benzoic acid, 3-bromo,3-bromo-benzoic acid,benzoic acid, m-bromo,unii-7dct3a73lx,7dct3a73lx,3-bromo benzene acid,3-bromobenzoicacid,4-fluorobenzoicacid
11456
3-bromobenzoic acid
C1=CC(=CC(=C1)Br)C(=O)O
585-76-2
C7H5BrO2
201.02
MFCD00002487
VOIZNVUXCQLQHS-UHFFFAOYSA-N
m-bromobenzoic acid,3-brombenzoic acid,benzoic acid, 3-bromo,3-bromo-benzoic acid,benzoic acid, m-bromo,unii-7dct3a73lx,7dct3a73lx,3-bromo benzene acid,3-bromobenzoicacid,4-fluorobenzoicacid
11456
3-bromobenzoic acid
C1=CC(=CC(=C1)Br)C(=O)O
Melting Point 154°C to 158°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 100g
Color White to Beige
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Linear Formula BrC6H4CO2H
Beilstein 09, 349
Solubility Information Solubility in water: 0.54g/L (32°C)
Formula Weight 201.02
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 3-Bromobenzoic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-562-3