Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobenzyl alcohol, 99%

Catalog No. AAA1092906
Click to view available options
Quantity:
5 g

CAS: 15852-73-0 | C7H7BrO | 187.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15852-73-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00004629
InChI Key FSWNRRSWFBXQCL-UHFFFAOYSA-N
Synonym 3-bromobenzyl alcohol, 3-bromophenyl methanol, m-bromobenzyl alcohol, benzenemethanol, 3-bromo, 3-bromobenzylalcohol, 3-bromophenyl methan-1-ol, 3-bromo-benzenemethanol, m-bromobenzylalcohol, 3-bromo benzylalcohol, pubchem3747
PubChem CID 85141
IUPAC Name (3-bromophenyl)methanol
SMILES C1=CC(=CC(=C1)Br)CO

Specifications

Density 1.56
Boiling Point 165°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.584
Quantity 5 g
Beilstein 2242512
Formula Weight 187.04
Percent Purity 99%
Chemical Name or Material 3-Bromobenzyl alcohol
Hazard Category H302+H332-H315-H319
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 239-975-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.