Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. B128725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B128725G 25 g
B12875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B128725G Supplier TCI America Supplier No. B128725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15852-73-0
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00004629
InChI Key FSWNRRSWFBXQCL-UHFFFAOYSA-N
Synonym 3-bromobenzyl alcohol, 3-bromophenyl methanol, m-bromobenzyl alcohol, benzenemethanol, 3-bromo, 3-bromobenzylalcohol, 3-bromophenyl methan-1-ol, 3-bromo-benzenemethanol, m-bromobenzylalcohol, 3-bromo benzylalcohol, pubchem3747
PubChem CID 85141
IUPAC Name (3-bromophenyl)methanol
SMILES C1=CC(=CC(=C1)Br)CO

Specifications

Color Yellow
Boiling Point 165°C
Quantity 25 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.