Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromobiphenyl, 99%

Click to view available options
Quantity:
1g
5g
25g
2113-57-7
C12H9Br
233.108
MFCD00000082
USYQKCQEVBFJRP-UHFFFAOYSA-N
3-bromobiphenyl,3-bromo-1,1'-biphenyl,1,1'-biphenyl, 3-bromo,3-bromo-biphenyl,m-bromobiphenyl,biphenyl, 3-bromo,3-bromodiphenyl,unii-o7i53k56s8,ccris 5889,3-brombiphenyl
16449
1-bromo-3-phenylbenzene
C1=CC=C(C=C1)C2=CC(=CC=C2)Br
2113-57-7
C12H9Br
233.108
MFCD00000082
USYQKCQEVBFJRP-UHFFFAOYSA-N
3-bromobiphenyl,3-bromo-1,1'-biphenyl,1,1'-biphenyl, 3-bromo,3-bromo-biphenyl,m-bromobiphenyl,biphenyl, 3-bromo,3-bromodiphenyl,unii-o7i53k56s8,ccris 5889,3-brombiphenyl
16449
1-bromo-3-phenylbenzene
C1=CC=C(C=C1)C2=CC(=CC=C2)Br
Applications
3-Bromobiphenyl is utilized as an electrochemical immunosensor based on poly(dopamine) coated gold nanocluster for brominated flame retardants.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Boiling Point 301°C
Flash Point >110°C (230°F)
Quantity 1g
Beilstein 2043191
Formula Weight 233.12
Percent Purity 99%
Chemical Name or Material 3-Bromobiphenyl

RTECSNumber : DV1750000

TSCA : TSCA