Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromobiphenyl, 99%

Catalog No. p-7025973
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 2113-57-7 | C12H9Br | 233.11 g/mol

3-Bromobiphenyl is utilized as an electrochemical immunosensor based on poly(dopamine) coated gold nanocluster for brominated flame retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromobiphenyl is utilized as an electrochemical immunosensor based on poly(dopamine) coated gold nanocluster for brominated flame retardants.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2113-57-7
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.11
MDL Number MFCD00000082
InChI Key USYQKCQEVBFJRP-UHFFFAOYSA-N
Synonym 3-bromobiphenyl, 3-bromo-1,1'-biphenyl, 1,1'-biphenyl, 3-bromo, 3-bromo-biphenyl, m-bromobiphenyl, biphenyl, 3-bromo, 3-bromodiphenyl, unii-o7i53k56s8, ccris 5889, 3-brombiphenyl
PubChem CID 16449
IUPAC Name 1-bromo-3-phenylbenzene
SMILES BrC1=CC=CC(=C1)C1=CC=CC=C1

Specifications

Density 1.397
Boiling Point 301°C
Flash Point >110°C (230°F)
Refractive Index 1.641
Quantity 1 g
Beilstein 2043191
Solubility Information Immiscible with water.
Formula Weight 233.12
Percent Purity 99%
Chemical Name or Material 3-Bromobiphenyl
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-304-9
RTECSNumber DV1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.