Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromocyclohexene 94.0+%, TCI America™
SDP

Catalog No. B094325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B094325ML 25 mL
B09435ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B094325ML Supplier TCI America Supplier No. B094325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1521-51-3
Molecular Formula C6H9Br
Molecular Weight (g/mol) 161.042
MDL Number MFCD00013775
InChI Key AJKDUJRRWLQXHM-UHFFFAOYSA-N
Synonym 3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene
PubChem CID 137057
IUPAC Name 3-bromocyclohexene
SMILES C1CC=CC(C1)Br

Specifications

Boiling Point 82°C
Quantity 25 mL
UN Number 1993
Formula Weight 161.04
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromocyclohexene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.