Learn More
3-Bromofuran, 97%, stab. with 0.5% calcium carbonate
Description
3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.
Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.

Specifications
Specifications
Density | 1.635 |
Boiling Point | 102°C to 104°C |
Flash Point | 3°C (37°F) |
Refractive Index | 1.496 |
Quantity | 5 g |
UN Number | UN1993 |
Beilstein | 105337 |
Solubility Information | Soluble in chloroform,ethyl acetate. Insoluble in water. |
Formula Weight | 146.98 |
Percent Purity | 97% |
Show More |
Safety and Handling
Hazard Category | H225-H315-H319-H335 |
Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
EINECSNumber | 244-744-6 |
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.