Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. I05245G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 2131-59-1
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD00004803
InChI Key ZMGMGHNOACSMQN-UHFFFAOYSA-N
Synonym 3-bromophenyl isothiocyanate, m-bromophenyl isothiocyanate, benzene, 1-bromo-3-isothiocyanato, isothiocyanic acid, m-bromophenyl ester, 3-bromophenylisothiocyanate, 1-bromo-3-isothiocyanato-benzene, acmc-1chhg, m-bromphenylisothiocyanat, zmgmghnoacsmqn-uhfffaoysa, #
PubChem CID 16481
IUPAC Name 1-bromo-3-isothiocyanatobenzene
SMILES C1=CC(=CC(=C1)Br)N=C=S

Specifications

Color White
Boiling Point 256°C
Quantity 5 g
UN Number 1759
Formula Weight 214.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromophenyl Isothiocyanate
RTECSNumber NX8350000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.