Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenylacetone, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
21906-32-1
C9H9BrO
213.08
MFCD00210400
FQIZFAJMBXZVOL-UHFFFAOYSA-N
1-3-bromophenyl propan-2-one,3-bromophenylacetone,1-3-bromophenyl-2-propanone,1-3-bromophenyl acetone,2-propanone, 1-3-bromophenyl,3-bromophenyl acetone,ksc492m0h,1-3-bromo-phenyl propan-2-one
2734093
1-(3-bromophenyl)propan-2-one
CC(=O)CC1=CC(=CC=C1)Br
21906-32-1
C9H9BrO
213.08
MFCD00210400
FQIZFAJMBXZVOL-UHFFFAOYSA-N
1-3-bromophenyl propan-2-one,3-bromophenylacetone,1-3-bromophenyl-2-propanone,1-3-bromophenyl acetone,2-propanone, 1-3-bromophenyl,3-bromophenyl acetone,ksc492m0h,1-3-bromo-phenyl propan-2-one
2734093
1-(3-bromophenyl)propan-2-one
CC(=O)CC1=CC(=CC=C1)Br
Refractive Index 1.5570 to 1.56
Packaging Glass bottle
Quantity 5g
Color Yellow
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Formula Weight 213.08
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material 3-Bromophenylacetone
Show More Show Less