Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromophenylacetonitrile, 97%

Catalog No. AAB2018914
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2018914 25 g
AAB2018906 5 g
AAB2018922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2018914 Supplier Thermo Scientific Chemicals Supplier No. B2018914
Add to Cart

CAS: 31938-07-5 | C8H6BrN | 196.047 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31938-07-5
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00001906
InChI Key UUZYFBXKWIQKTF-UHFFFAOYSA-N
Synonym 3-bromophenylacetonitrile, 3-bromobenzyl cyanide, 2-3-bromophenyl acetonitrile, m-bromobenzyl cyanide, benzeneacetonitrile, 3-bromo, 3-bromobenzylcyanide, 3-bromobenzeneacetonitrile, 3-bromophenyl acetonitrile, m-bromophenylacetonitrile, 2-3-bromophenyl ethanenitrile
PubChem CID 36023
IUPAC Name 2-(3-bromophenyl)acetonitrile
SMILES C1=CC(=CC(=C1)Br)CC#N

Specifications

Melting Point 25°C to 28°C
Boiling Point 275°C
Flash Point 126°C (259°F)
Refractive Index 1.57
Quantity 25 g
UN Number UN3449
Beilstein 2355439
Formula Weight 196.05
Percent Purity 97%
Chemical Name or Material 3-Bromophenylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H300-H310-H330-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: BROMOBENZYL CYANIDES, SOLID
EINECSNumber 250-867-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.